Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY001125379

Key Organics / BIONET

(6-bromo-2H-1,3-benzodioxol-5-yl)methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
231.05
MDL number:

mp

90 - 92

Iupac Name

(6-bromo-1,3-benzodioxol-5-yl)methanol

Inchi Code

1S/C8H7BrO3/c9-6-2-8-7(11-4-12-8)1-5(6)3-10/h1-2,10H,3-4H2

InChI key

XYYBAJXSNPIXTC-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/10P-113

Melting Point

90 - 92

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/10P-113


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service