Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY001123865

Key Organics / BIONET

4-(pyrrolidin-1-yl)aniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
162.23
MDL number:

bp

330.8C @ 760mmHg

mp

32 - 34C

Iupac Name

4-(1-pyrrolidinyl)phenylamine

Inchi Code

1S/C10H14N2/c11-9-3-5-10(6-4-9)12-7-1-2-8-12/h3-6H,1-2,7-8,11H2

InChI key

URAARCWOADCWLA-UHFFFAOYSA-N

storage temp.

Store under inert gas at 4C

Purity

95%

Country of Origin

GB

Physical Form

Low Melting Solid

Product Link

https://www.keyorganics.net/sku/10G-314S

Boiling point

330.8C @ 760mmHg

Melting Point

32 - 34C

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/10G-314S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service