Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JWPH3249EB2B

J & W Pharmlab LLC

1-Phenyl-1,2,3,4-tetrahydro-quinolin-4-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
224.31
MDL number:

Iupac Name

1-phenyl-1,2,3,4-tetrahydro-4-quinolinamine

Inchi Code

1S/C15H16N2/c16-14-10-11-17(12-6-2-1-3-7-12)15-9-5-4-8-13(14)15/h1-9,14H,10-11,16H2

InChI key

JDIQRZNUOJQPCT-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Yellow Liquid

Product Link

https://www.jwpharmlab.com/home/product/80R0273

MSDS

https://www.jwpharmlab.com/home/product/80R0273


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service