Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JWPH3249DBF6

J & W Pharmlab LLC

1-Benzyl-2,6-dimethyl-piperidin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
217.31
MDL number:

Iupac Name

1-benzyl-2,6-dimethyl-4-piperidinone

Inchi Code

1S/C14H19NO/c1-11-8-14(16)9-12(2)15(11)10-13-6-4-3-5-7-13/h3-7,11-12H,8-10H2,1-2H3

InChI key

RZABKAGQMPNUFJ-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Light Yellow Solid

Product Link

https://www.jwpharmlab.com/home/product/60R0856

MSDS

https://www.jwpharmlab.com/home/product/60R0856


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service