Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JWPH3249C6B4

J & W Pharmlab LLC

1-Aminomethyl-cyclopentanecarboxylic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
199.29
MDL number:

Iupac Name

tert-butyl 1-(aminomethyl)cyclopentanecarboxylate

Inchi Code

1S/C11H21NO2/c1-10(2,3)14-9(13)11(8-12)6-4-5-7-11/h4-8,12H2,1-3H3

InChI key

CMWYGCVPORHJMO-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Red Liquid

Product Link

https://www.jwpharmlab.com/home/product/15R0210

MSDS

https://www.jwpharmlab.com/home/product/15R0210


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service