Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ELGH9602CB7B

Exelgen Discovery Ltd.

2,5-dibutylfuran

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
180.29
MDL number:

Iupac Name

2,5-dibutylfuran

Inchi Code

1S/C12H20O/c1-3-5-7-11-9-10-12(13-11)8-6-4-2/h9-10H,3-8H2,1-2H3

InChI key

XTBSCDXJHDVPLX-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

liquid

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service