Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH98BAA85B

Enamine

(3-aminophenyl)phosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
173.11
MDL number:

mp

288-290

Iupac Name

3-aminophenylphosphonic acid

Inchi Code

1S/C6H8NO3P/c7-5-2-1-3-6(4-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)

InChI key

MZZQBSHNCYWSTL-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-6770389

Melting Point

288-290

MSDS

https://www.enaminestore.com/catalog/EN300-6770389


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service