Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH984B76AE

Enamine

2-amino-4-(2,5-dichlorophenyl)thiophene-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
269.15
MDL number:

Iupac Name

2-amino-4-(2,5-dichlorophenyl)-3-thiophenecarbonitrile

Inchi Code

1S/C11H6Cl2N2S/c12-6-1-2-10(13)7(3-6)9-5-16-11(15)8(9)4-14/h1-3,5H,15H2

InChI key

JTUYPUFMYQJTIR-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-6503621

MSDS

https://www.enaminestore.com/catalog/EN300-6503621


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service