Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH961CEA97

Enamine

N-(3,5-dichlorophenyl)hydroxylamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
178.02
MDL number:

mp

85-87

Iupac Name

N-(3,5-dichlorophenyl)hydroxylamine

Inchi Code

1S/C6H5Cl2NO/c7-4-1-5(8)3-6(2-4)9-10/h1-3,9-10H

InChI key

LBFDAACXWGTMIX-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-7500274

Melting Point

85-87

MSDS

https://www.enaminestore.com/catalog/EN300-7500274


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service