Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH961CD7EB

Enamine

ethyl 5-{[(tert-butoxy)carbonyl]amino}-2,2-difluoro-4-iodopentanoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
407.2
MDL number:

Iupac Name

ethyl 5-((tert-butoxycarbonyl)amino)-2,2-difluoro-4-iodopentanoate

Inchi Code

1S/C12H20F2INO4/c1-5-19-9(17)12(13,14)6-8(15)7-16-10(18)20-11(2,3)4/h8H,5-7H2,1-4H3,(H,16,18)

InChI key

VYTDLKOMTMCGQT-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-27694424

MSDS

https://www.enaminestore.com/catalog/EN300-27694424


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service