Skip to Content
MilliporeSigma

ENAH942F3032

Enamine

methyl 2-[3-(difluoromethoxy)phenyl]-2-oxoacetate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
230.17
MDL number:

Pricing and availability is not currently available.

Iupac Name

methyl 2-(3-(difluoromethoxy)phenyl)-2-oxoacetate

Inchi Code

1S/C10H8F2O4/c1-15-9(14)8(13)6-3-2-4-7(5-6)16-10(11)12/h2-5,10H,1H3

InChI key

DDOKZLKZCYHZPI-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB4502936SAB4503801SAB3500372
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

mol wt

antigen 76 kDa

mol wt

antigen 76 kDa

mol wt

antigen 76 kDa

mol wt

predicted mol wt 76 kDa

species reactivity

rat, human, mouse

species reactivity

mouse, human, rat

species reactivity

mouse, human, rat

species reactivity

human, mouse

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-23988587

MSDS

https://www.enaminestore.com/catalog/EN300-23988587


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service