Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH90822EDE

Enamine

methyl 5-(2-methoxyethyl)-1,2-oxazole-3-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€106.00

€106.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€106.00

About This Item

CAS Number:
Molecular Weight:
185.18
MDL number:

€106.00


Please contact Customer Service for Availability

Iupac Name

methyl 5-(2-methoxyethyl)isoxazole-3-carboxylate

Inchi Code

1S/C8H11NO4/c1-11-4-3-6-5-7(9-13-6)8(10)12-2/h5H,3-4H2,1-2H3

InChI key

MANJAXONNURCEP-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
961000299610003296100030
manufacturer/tradename

Vantage

manufacturer/tradename

Vantage

manufacturer/tradename

Vantage

manufacturer/tradename

Vantage

technique(s)

protein purification: suitable

technique(s)

protein purification: suitable

technique(s)

protein purification: suitable

technique(s)

protein purification: suitable

diam.

2.2 cm (0.9 in.)

diam.

1.6 cm (0.6 in.)

diam.

4.4 cm (1.7 in.)

diam.

2.2 cm (0.9 in.)

Quality Level

SP1

Quality Level

SP1

Quality Level

SP1

Quality Level

SP1

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-7445243

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-7445243


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service