Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH65927B1B

Enamine

1H-pyrazole-5-sulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
150.13
MDL number:

mp

125-126

Iupac Name

1H-pyrazole-5-sulfonyl fluoride

Inchi Code

1S/C3H3FN2O2S/c4-9(7,8)3-1-2-5-6-3/h1-2H,(H,5,6)

InChI key

DDADMFYLUVJHKR-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-256911

Melting Point

125-126

MSDS

https://www.enaminestore.com/catalog/EN300-256911


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service