Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH5802F47B

Enamine

3-cyclopropyl-1,2-oxazole-5-sulfonyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$410.80
100 MG
$611.00
250 MG
$873.60
500 MG
$1,375.40
1 G
$1,764.10
2.5 G
$3,458.00
5 G
$5,115.50

Vendor SKU: EN300-6740583-50MG

$410.80


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
50 MG
$410.80
100 MG
$611.00
250 MG
$873.60
500 MG
$1,375.40
1 G
$1,764.10
2.5 G
$3,458.00
5 G
$5,115.50

About This Item

CAS Number:
Molecular Weight:
207.64
MDL number:

Vendor SKU: EN300-6740583-50MG

$410.80


Ships in 7 days - from Aldrich Partner.

Iupac Name

3-cyclopropylisoxazole-5-sulfonyl chloride

Inchi Code

1S/C6H6ClNO3S/c7-12(9,10)6-3-5(8-11-6)4-1-2-4/h3-4H,1-2H2

InChI key

JRBGTKYWJCIGHD-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-6740583

MSDS

https://www.enaminestore.com/catalog/EN300-6740583


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service