ENAH38A2CF50
Enamine
4-hydroxy-N-propylbenzene-1-sulfonamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
82-84
Iupac Name
4-hydroxy-N-propylbenzenesulfonamide
Inchi Code
1S/C9H13NO3S/c1-2-7-10-14(12,13)9-5-3-8(11)4-6-9/h3-6,10-11H,2,7H2,1H3
InChI key
WUUUOCMXAGTQFV-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-6759775
Melting Point
82-84
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-6759775
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service