Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH31551C0A

Enamine

(E)-2-(4-chlorophenyl)ethene-1-sulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
220.65
MDL number:

mp

127-128

Iupac Name

(E)-2-(4-chlorophenyl)ethene-1-sulfonyl fluoride

Inchi Code

1S/C8H6ClFO2S/c9-8-3-1-7(2-4-8)5-6-13(10,11)12/h1-6H/b6-5+

InChI key

UVUNWXQELONMGK-AATRIKPKSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-6732265

Melting Point

127-128

pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-6732265


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service