Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH304A75F0

Enamine

3-amino-3-(3,5-dimethylphenyl)propanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,001.00
2.5 G
$1,963.00
5 G
$2,905.50
10 G
$4,309.50

Vendor SKU: EN300-816582-1G

$1,001.00


Ships in 30 days - from Aldrich Partner.


Select a Size

Change View
1 G
$1,001.00
2.5 G
$1,963.00
5 G
$2,905.50
10 G
$4,309.50

About This Item

CAS Number:
Molecular Weight:
193.25
MDL number:

Vendor SKU: EN300-816582-1G

$1,001.00


Ships in 30 days - from Aldrich Partner.

Iupac Name

3-(3,5-dimethylphenyl)-beta-alanine

Inchi Code

1S/C11H15NO2/c1-7-3-8(2)5-9(4-7)10(12)6-11(13)14/h3-5,10H,6,12H2,1-2H3,(H,13,14)

InChI key

YBIJIOITROSVBG-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-816582

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service