Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH304A311C

Enamine

1-cyclopentyl-1H-1,2,3-triazol-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
152.2
MDL number:

Iupac Name

1-cyclopentyl-1H-1,2,3-triazol-4-amine

Inchi Code

1S/C7H12N4/c8-7-5-11(10-9-7)6-3-1-2-4-6/h5-6H,1-4,8H2

InChI key

DNASZFNYXLYNPB-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-783944

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service