Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH30488C8D

Enamine

4-ethyl-6-methoxy-1H,2H,3H,4H-pyrido[2,3-b]pyrazine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,280.50
2.5 G
$2,509.00
5 G
$3,714.10
10 G
$5,508.10

Vendor SKU: EN300-694939-1G

$1,280.50


Ships in 30 days - from Aldrich Partner.


Select a Size

Change View
1 G
$1,280.50
2.5 G
$2,509.00
5 G
$3,714.10
10 G
$5,508.10

About This Item

CAS Number:
Molecular Weight:
193.25
MDL number:

Vendor SKU: EN300-694939-1G

$1,280.50


Ships in 30 days - from Aldrich Partner.

Iupac Name

4-ethyl-6-methoxy-1,2,3,4-tetrahydropyrido[2,3-b]pyrazine

Inchi Code

1S/C10H15N3O/c1-3-13-7-6-11-8-4-5-9(14-2)12-10(8)13/h4-5,11H,3,6-7H2,1-2H3

InChI key

RLWNFNBUIKOPIQ-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-694939

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service