ENAH3046C7E6
Enamine
1,5,5-trimethyl-6',7'-dihydro-5'H-spiro[pyrrolidine-3,4'-thieno[3,2-c]pyridine]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
236.38
Iupac Name
1,5,5-trimethyl-6',7'-dihydro-5'H-spiro[pyrrolidine-3,4'-thieno[3,2-c]pyridine]
Inchi Code
1S/C13H20N2S/c1-12(2)8-13(9-15(12)3)10-5-7-16-11(10)4-6-14-13/h5,7,14H,4,6,8-9H2,1-3H3
InChI key
LLKBWURMELQMSO-UHFFFAOYSA-N
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-1076864
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service