Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH304647F9

Enamine

1-(2-phenylcyclopropyl)ethan-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$686.40
2.5 G
$1,344.20
5 G
$1,989.00
10 G
$2,951.00

Vendor SKU: EN300-52579-1G

$686.40


Ships in 30 days - from Aldrich Partner.


Select a Size

Change View
1 G
$686.40
2.5 G
$1,344.20
5 G
$1,989.00
10 G
$2,951.00

About This Item

CAS Number:
Molecular Weight:
160.22
MDL number:

Vendor SKU: EN300-52579-1G

$686.40


Ships in 30 days - from Aldrich Partner.

Iupac Name

1-(2-phenylcyclopropyl)ethanone

Inchi Code

1S/C11H12O/c1-8(12)10-7-11(10)9-5-3-2-4-6-9/h2-6,10-11H,7H2,1H3

InChI key

LFKRVDCVJRDOBG-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-52579

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service