Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH304578CE

Enamine

propyl 2-hydroxy-2-phenylacetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$872.30
2.5 G
$1,708.20
5 G
$2,528.50
10 G
$3,749.20

Vendor SKU: EN300-125424-1G

$872.30


Ships in 30 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 G
$872.30
2.5 G
$1,708.20
5 G
$2,528.50
10 G
$3,749.20

About This Item

CAS Number:
Molecular Weight:
194.23
MDL number:

Vendor SKU: EN300-125424-1G

$872.30


Ships in 30 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

propyl hydroxy(phenyl)acetate

Inchi Code

1S/C11H14O3/c1-2-8-14-11(13)10(12)9-6-4-3-5-7-9/h3-7,10,12H,2,8H2,1H3

InChI key

MKZJVQWJKDMCKK-UHFFFAOYSA-N

Purity

85

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-125424

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service