Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH2C55828D

Enamine

1-hydroxypyrrolidin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
101.11
MDL number:

mp

68-69

Iupac Name

1-hydroxypyrrolidin-2-one

Inchi Code

1S/C4H7NO2/c6-4-2-1-3-5(4)7/h7H,1-3H2

InChI key

JQBCKRZQAMELAD-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-6482819

Melting Point

68-69

MSDS

https://www.enaminestore.com/catalog/EN300-6482819


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service