Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH11E37EF4

Enamine

1-(4-chloro-2-fluorophenyl)cyclopropane-1-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
214.62
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

1-(4-chloro-2-fluorophenyl)cyclopropanecarboxylic acid

Inchi Code

1S/C10H8ClFO2/c11-6-1-2-7(8(12)5-6)10(3-4-10)9(13)14/h1-2,5H,3-4H2,(H,13,14)

InChI key

REFBTNOOKKSXRG-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-1984483

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-1984483


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service