Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH0424B4C6

Enamine

5-fluoro-1-(4-phenylphenyl)-1H-pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
238.26
MDL number:

mp

59-60

Iupac Name

1-([1,1'-biphenyl]-4-yl)-5-fluoro-1H-pyrazole

Inchi Code

1S/C15H11FN2/c16-15-10-11-17-18(15)14-8-6-13(7-9-14)12-4-2-1-3-5-12/h1-11H

InChI key

KYPIZDCRZFSHCK-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1272035

Melting Point

59-60

MSDS

https://www.enaminestore.com/catalog/EN300-1272035


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service