Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH03A018AF

Enamine

(dimethylcarbamoyl)methanesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
185.63
MDL number:

Iupac Name

2-(dimethylamino)-2-oxoethane-1-sulfonyl chloride

Inchi Code

1S/C4H8ClNO3S/c1-6(2)4(7)3-10(5,8)9/h3H2,1-2H3

InChI key

QCOLUTLBSUAPTR-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-286537

MSDS

https://www.enaminestore.com/catalog/EN300-286537


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service