Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH03A003B2

Enamine

3,3-dimethylcyclobutane-1-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
112.17
MDL number:

Iupac Name

3,3-dimethylcyclobutane-1-carbaldehyde

Inchi Code

1S/C7H12O/c1-7(2)3-6(4-7)5-8/h5-6H,3-4H2,1-2H3

InChI key

HIPQXNTZSDKPRC-UHFFFAOYSA-N

storage temp.

-10

Purity

91

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-1081155

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service