Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA964469159

Enamine

methyl 4-{[3-(2,4-dihydroxyphenyl)-1H-pyrazol-4-yl]oxy}benzoate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
326.31
MDL number:

Iupac Name

methyl 4-{[3-(2,4-dihydroxyphenyl)-1H-pyrazol-4-yl]oxy}benzoate

Inchi Code

1S/C17H14N2O5/c1-23-17(22)10-2-5-12(6-3-10)24-15-9-18-19-16(15)13-7-4-11(20)8-14(13)21/h2-9,20-21H,1H3,(H,18,19)

InChI key

NKLDVDYKUHYJNO-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-395900

MSDS

https://www.enaminestore.com/catalog/EN300-395900


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service