Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA964468826

Enamine

4-chloro-6-(oxan-4-yloxy)pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
214.65
MDL number:

Iupac Name

6-chloro-4-pyrimidinyl tetrahydro-2H-pyran-4-yl ether

Inchi Code

1S/C9H11ClN2O2/c10-8-5-9(12-6-11-8)14-7-1-3-13-4-2-7/h5-7H,1-4H2

InChI key

JBBCTAXBMXZBPK-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-655519

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-655519


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service