Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA964468206

Enamine

2-methyl-4-(trifluoromethyl)furan-3-carbonyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$335.40
250 MG
$477.10
500 MG
$752.70
1 G
$965.90
2.5 G
$1,890.20
5 G
$2,797.60

Vendor SKU: EN300-384460-100MG

$335.40


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$335.40
250 MG
$477.10
500 MG
$752.70
1 G
$965.90
2.5 G
$1,890.20
5 G
$2,797.60

About This Item

CAS Number:
Molecular Weight:
212.56
MDL number:

Vendor SKU: EN300-384460-100MG

$335.40


Ships in 7 days - from Aldrich Partner.

Iupac Name

2-methyl-4-(trifluoromethyl)furan-3-carbonyl chloride

Inchi Code

1S/C7H4ClF3O2/c1-3-5(6(8)12)4(2-13-3)7(9,10)11/h2H,1H3

InChI key

STLAONYMTMTOAO-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-384460

MSDS

https://www.enaminestore.com/catalog/EN300-384460


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service