Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA497682981

Enamine

2-(5-amino-2-oxo-1,2-dihydropyridin-1-yl)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$196.30
250 MG
$280.80
500 MG
$529.10
1 G
$686.40
2.5 G
$1,344.20
5 G
$1,989.00

Vendor SKU: EN300-58400-100MG

$196.30


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$196.30
250 MG
$280.80
500 MG
$529.10
1 G
$686.40
2.5 G
$1,344.20
5 G
$1,989.00

About This Item

CAS Number:
Molecular Weight:
167.17
MDL number:

Vendor SKU: EN300-58400-100MG

$196.30


Ships in 7 days - from Aldrich Partner.

Iupac Name

2-(5-amino-2-oxo-1(2H)-pyridinyl)acetamide

Inchi Code

1S/C7H9N3O2/c8-5-1-2-7(12)10(3-5)4-6(9)11/h1-3H,4,8H2,(H2,9,11)

InChI key

SFALCUCWFPEYPA-UHFFFAOYSA-N

storage temp.

-10

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-58400

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-58400


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service