Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA490046884

Enamine

N-methoxy-N-methyl-4-oxopentanamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$196.30
250 MG
$280.80
500 MG
$529.10
1 G
$686.40
2.5 G
$1,344.20
5 G
$1,989.00
10 G
$2,951.00

Vendor SKU: EN300-245310-100MG

$196.30


Availability
Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$196.30
250 MG
$280.80
500 MG
$529.10
1 G
$686.40
2.5 G
$1,344.20
5 G
$1,989.00
10 G
$2,951.00

About This Item

CAS Number:
Molecular Weight:
159.19
MDL number:

Vendor SKU: EN300-245310-100MG

$196.30


Availability
Ships in 7 days - from Aldrich Partner.

Iupac Name

N-methoxy-N-methyl-4-oxopentanamide

Inchi Code

1S/C7H13NO3/c1-6(9)4-5-7(10)8(2)11-3/h4-5H2,1-3H3

InChI key

XOHQJZCCBJVTED-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-245310

MSDS

https://www.enaminestore.com/catalog/EN300-245310


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service