Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA458426627

Enamine

ethyl N-(4-bromo-3-oxo-1-phenylbutan-2-yl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
314.18
MDL number:

Iupac Name

ethyl (4-bromo-3-oxo-1-phenylbutan-2-yl)carbamate

Inchi Code

1S/C13H16BrNO3/c1-2-18-13(17)15-11(12(16)9-14)8-10-6-4-3-5-7-10/h3-7,11H,2,8-9H2,1H3,(H,15,17)

InChI key

PWMOVBGWHOEIOS-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-226905

MSDS

https://www.enaminestore.com/catalog/EN300-226905


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service