ENA448968437
Enamine
5-fluoro-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
155-160
Iupac Name
5-fluoro-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole
Inchi Code
1S/C13H13FN2/c14-10-1-2-13-11(7-10)12(8-16-13)9-3-5-15-6-4-9/h1-3,7-8,15-16H,4-6H2
InChI key
NOBJPJHWOFGLFA-UHFFFAOYSA-N
storage temp.
4
Purity
93
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-211881
Melting Point
155-160
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-211881
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service