ENA424046252
Enamine
tert-butyl 4-(2-oxoimidazolidin-1-yl)piperidine-1-carboxylate
About This Item
Iupac Name
tert-butyl 4-(2-oxo-1-imidazolidinyl)-1-piperidinecarboxylate
Inchi Code
1S/C13H23N3O3/c1-13(2,3)19-12(18)15-7-4-10(5-8-15)16-9-6-14-11(16)17/h10H,4-9H2,1-3H3,(H,14,17)
InChI key
NPTZWFCKLAFHIL-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-94956
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service