Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA424046092

Enamine

iodomethyl 2-methylpropanoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
228.03
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

iodomethyl isobutyrate

Inchi Code

1S/C5H9IO2/c1-4(2)5(7)8-3-6/h4H,3H2,1-2H3

InChI key

KWIBRJGXWFDQNU-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-177240

MSDS

https://www.enaminestore.com/catalog/EN300-177240


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service