ENA423207815
Enamine
N-[(3-fluorophenyl)methyl]cyclobutanamine
Select a Size
Vendor SKU: EN300-168379-100MG
$313.30
Select a Size
About This Item
Vendor SKU: EN300-168379-100MG
$313.30
Iupac Name
N-(3-fluorobenzyl)cyclobutanamine
Inchi Code
1S/C11H14FN/c12-10-4-1-3-9(7-10)8-13-11-5-2-6-11/h1,3-4,7,11,13H,2,5-6,8H2
InChI key
QCXOXTBHOBCJJJ-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-168379
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service