ENA413170878
Enamine
3-aminocyclopentane-1-carbonitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
3-aminocyclopentanecarbonitrile
Inchi Code
1S/C6H10N2/c7-4-5-1-2-6(8)3-5/h5-6H,1-3,8H2
InChI key
PKTNSCWXNMMDQK-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-152064
MSDS
https://www.enaminestore.com/catalog/EN300-152064
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service