ENA410542398
Enamine
(1-amino-2-ethoxy-2-oxoethylidene)amino propanoate
Select a Size
Vendor SKU: EN300-138981-100MG
$352.30
Availability
Select a Size
About This Item
Vendor SKU: EN300-138981-100MG
$352.30
Availability
Iupac Name
ethyl (Z)-2-amino-2-((propionyloxy)imino)acetate
Inchi Code
1S/C7H12N2O4/c1-3-5(10)13-9-6(8)7(11)12-4-2/h3-4H2,1-2H3,(H2,8,9)
InChI key
BZKIBWGEJBPOEC-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-138981
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service