Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA402267588

Enamine

thiane-2-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
127.21
MDL number:

mp

29-30

Iupac Name

tetrahydro-2H-thiopyran-2-carbonitrile

Inchi Code

1S/C6H9NS/c7-5-6-3-1-2-4-8-6/h6H,1-4H2

InChI key

ZUBIQKKIDXWWHU-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-107696

Melting Point

29-30

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-107696


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service