Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA394767743

Enamine

2-(3,5-dimethyl-1H-pyrazol-4-yl)azepane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
193.29
MDL number:

Iupac Name

2-(3,5-dimethyl-1H-pyrazol-4-yl)azepane

Inchi Code

1S/C11H19N3/c1-8-11(9(2)14-13-8)10-6-4-3-5-7-12-10/h10,12H,3-7H2,1-2H3,(H,13,14)

InChI key

QWQQOCFYULMGMI-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

oil

Product Link

https://www.enaminestore.com/catalog/EN300-123141

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-123141


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service