ENA394767711
Enamine
2-bromo-N-(3-ethynylphenyl)acetamide
Select a Size
Vendor SKU: EN300-129215-100MG
$187.20
Select a Size
About This Item
Vendor SKU: EN300-129215-100MG
$187.20
mp
120-125
Iupac Name
2-bromo-N-(3-ethynylphenyl)acetamide
Inchi Code
1S/C10H8BrNO/c1-2-8-4-3-5-9(6-8)12-10(13)7-11/h1,3-6H,7H2,(H,12,13)
Inchi Key
SDYFXLKHSLLUAK-UHFFFAOYSA-N
Purity
Country of Origin
Product Link
Melting Point
MSDS
https://www.enaminestore.com/catalog/EN300-129215
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service