ENA394766902
Enamine
N-[1-(2-chlorophenyl)-1-methoxypropan-2-yl]hydroxylamine
Select a Size
Vendor SKU: EN300-128098-100MG
$444.60
Select a Size
About This Item
Vendor SKU: EN300-128098-100MG
$444.60
Iupac Name
N-(1-(2-chlorophenyl)-1-methoxypropan-2-yl)hydroxylamine
Inchi Code
1S/C10H14ClNO2/c1-7(12-13)10(14-2)8-5-3-4-6-9(8)11/h3-7,10,12-13H,1-2H3
Inchi Key
RKXBNTAHBSGATO-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
MSDS
https://www.enaminestore.com/catalog/EN300-128098
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service