ENA358813159
Enamine
(4-aminobutan-2-yl)(benzyl)methylamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
N~3~-benzyl-N~3~-methyl-1,3-butanediamine
Inchi Code
1S/C12H20N2/c1-11(8-9-13)14(2)10-12-6-4-3-5-7-12/h3-7,11H,8-10,13H2,1-2H3
InChI key
HGDNYWBJWULGKT-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
liquid
Product Link
https://www.enaminestore.com/catalog/EN300-97022
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-97022
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service