Iupac Name
(6-methoxy-3-pyridinyl)methanol
Inchi Code
1S/C7H9NO2/c1-10-7-3-2-6(5-9)4-8-7/h2-4,9H,5H2,1H3
InChI key
YKTYUHMWNUVAAM-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
liquid
Product Link
https://www.enaminestore.com/catalog/EN300-91610
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-91610
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service