ENA341294713
Enamine
1-oxa-4-azaspiro[5.5]undecane hydrochloride
About This Item
Iupac Name
1-oxa-4-azaspiro[5.5]undecane hydrochloride
Inchi Code
1S/C9H17NO.ClH/c1-2-4-9(5-3-1)8-10-6-7-11-9;/h10H,1-8H2;1H
InChI key
GTFVSELRZMIVBL-UHFFFAOYSA-N
storage temp.
Salt Data
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-90263
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service