ENA321765991
Enamine
1-(1-methyl-1H-imidazol-2-yl)ethan-1-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-(1-methyl-1H-imidazol-2-yl)ethanone
Inchi Code
1S/C6H8N2O/c1-5(9)6-7-3-4-8(6)2/h3-4H,1-2H3
InChI key
IZVOHCMELOUFRA-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
liquid
Product Link
https://www.enaminestore.com/catalog/EN300-77828
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-77828
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service