Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA308584853

Enamine

(2,2-difluoroethyl)[(pyridin-3-yl)methyl]amine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$391.30
250 MG
$560.30

Vendor SKU: EN300-69758-100MG

$391.30


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$391.30
250 MG
$560.30

About This Item

CAS Number:
Molecular Weight:
172.18
MDL number:

Vendor SKU: EN300-69758-100MG

$391.30


Ships in 7 days - from Aldrich Partner.

Iupac Name

2,2-difluoro-N-(3-pyridinylmethyl)ethanamine

Inchi Code

1S/C8H10F2N2/c9-8(10)6-12-5-7-2-1-3-11-4-7/h1-4,8,12H,5-6H2

Inchi Key

IZQAMADABUKNJQ-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-69758

MSDS

https://www.enaminestore.com/catalog/EN300-69758


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service