Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA292596497

Enamine

1-[(5-chlorothiophen-2-yl)sulfonyl]-1,4-diazepane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
280.8
MDL number:

Iupac Name

1-[(5-chloro-2-thienyl)sulfonyl]-1,4-diazepane

Inchi Code

1S/C9H13ClN2O2S2/c10-8-2-3-9(15-8)16(13,14)12-6-1-4-11-5-7-12/h2-3,11H,1,4-7H2

InChI key

XUVLOCJEHKEAQH-UHFFFAOYSA-N

Purity

93

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-62202

MSDS

https://www.enaminestore.com/catalog/EN300-62202


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service