Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA181911205

Enamine

3-hydrazinyl-5-methyl-4-phenyl-4H-1,2,4-triazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
189.22
MDL number:

Iupac Name

3-hydrazino-5-methyl-4-phenyl-4H-1,2,4-triazole

Inchi Code

1S/C9H11N5/c1-7-12-13-9(11-10)14(7)8-5-3-2-4-6-8/h2-6H,10H2,1H3,(H,11,13)

InChI key

KJRVPEQUBAHPFW-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-37017

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-37017


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service